Learn More
Trajan Scientific and Medical™ Removable Needle for Thermo Scientific RSH Autosampler
Click to view available options
Needle Gauge:
23 G
26 G
Volume (Metric):
5 μL
10 μL
Length (Metric):
57 mm
85 mm
Diameter (Metric) Outer:
0.47 mm
0.63 mm
Unit Size:
Pack of 2
Description
- Replacement needle for use with Thermo Scientific RSH Autosampler
- Removable needle
- Cone Tip
Specifications
Specifications
| Needle Gauge | 26 G |
| Diameter (Metric) Outer | 0.47 mm |
| Length (Metric) | 57 mm |
| Product Type | Removable Needle |
| Quantity | 2/Pk. |
| Volume (Metric) | 10 μL |
| Needle Point Style | Cone |
| For Use With (Equipment) | Thermo RSH Autosampler |