Learn More
Renalase Antibody (1C11E8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-52554
 View more versions of this product
Product Code. 18237750
  • € 552.00 / 0.10mg
Description
Renalase Monoclonal specifically detects Renalase in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Renalase
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready
C10orf59chromosome 10 open reading frame 59, EC 1.4, FLJ11218, MAO-C, Monoamine oxidase-C, RENALASE, renalase, FAD-dependent amine oxidase
Mouse
37.8 kDa
0.1 mg
Lipid and Metabolism
55328
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry
1C11E8
Unconjugated
PBS with 0.05% Sodium Azide
RNLS
Purified recombinant fragment of human RNLS (AA: 68-242) expressed in E. Coli.
Protein A or G purified