missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Renalase Monoclonal Antibody (C5)

Mouse Monoclonal Antibody
€ 214.00 - € 518.00
Specifications
| Antigen | Renalase |
|---|---|
| Clone | C5 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17867112
![]() |
Invitrogen™
MA541668 |
200 μL |
€ 518.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17644066
|
Invitrogen™
MA543608 |
20 μL |
€ 214.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Renalase Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Renalase is a flavoprotein enzyme encoded by the RNLS gene, primarily expressed in the kidneys and heart where it plays a critical role in metabolizing catecholamines and catecholamine-like substances. It utilizes nicotinamide adenine dinucleotide (NADH) as a cofactor and operates through a superoxide-dependent mechanism, which is distinct from monoamine oxidase pathways. Renalase is considered a novel renal hormone that influences blood pressure and cardiac function by regulating circulating catecholamines. Its genetic variants have been associated with essential hypertension, highlighting its importance in cardiovascular health. In diabetic nephropathy, renalase exhibits protective effects, as demonstrated in genetically modified mouse models. These models have shown that renalase overexpression can mitigate renal damage, suggesting therapeutic potential. Renalase's expression is highest in the proximal tubules of the kidneys, which are densely innervated by sympathetic nerves, underscoring the connection between renalase and sympathetic nervous system activity. This enzyme's discovery has enriched our understanding of renal hormone regulation and its implications in diseases such as chronic kidney disease (CKD) and end-stage renal disease (ESRD).Specifications
| Renalase | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5U2W9, Q5VYX0 | |
| 361751, 55328 | |
| Recombinant protein Renalase. The antigen corresponds to amino acid range 18-342 of the target protein. | |
| Antibody |
| C5 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human, Rat | |
| Rnls | |
| 6530404N21Rik; AI452315; alpha-NAD(P)H oxidase/anomerase; AW060440; C10orf59; FLJ11218; HGNC:25641; MAO-C; mMAO-C; monoamine oxidase-C; Renalase; renalase, FAD dependent amine oxidase; renalase, FAD-dependent amine oxidase; RGD1309804; RNLS | |
| Rnls | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title