missing translation for 'onlineSavingsMsg'
Learn More

Renin Antibody (SR1820), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 338.00 - € 449.00

Specifications

Antigen Renin
Clone SR1820
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50
Applications Western Blot, Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18690985
View Documents
Novus Biologicals
NBP3-22259-100ul
100 μg
€ 449.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18673855
View Documents
Novus Biologicals
NBP3-22259-50ul
50 μg
€ 338.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Renin Monoclonal antibody specifically detects Renin in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications

Specifications

Renin
Western Blot 1:500-1:2000, Flow Cytometry 1:50
Monoclonal
Purified
RUO
Human
angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, Prorenin, renin, renin precursor, renal
A synthesized peptide derived from human Renin (Uniprot #: P00797)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1820
Western Blot, Flow Cytometry
Unconjugated
Rabbit
Cardiovascular Biology, metabolism
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
5972
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.