Learn More
Invitrogen™ Renin Monoclonal Antibody (411525)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524151
Description
This antibody detects the pro (aa 24-406) and mature (aa 67-406) forms of human Renin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Renin, recombinant human (rh) Cathepsin D, or rhCathepsin E is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.
Specifications
| Renin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P00797 | |
| Ren | |
| Mouse myeloma cell line NS0-derived recombinant human Renin Leu24-Arg406. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| 411525 | |
| Unconjugated | |
| Ren | |
| angiotensin-forming enzyme; angiotensinogenase; aspartyl-protease; D19352; EC 3.4.23; EC 3.4.23.15; FLJ10761; HNFJ2; kidney renin; RATRENAA; Ren; Ren1; Ren-1; Ren1c; Ren1d; Ren-A; RENAA; Renin; renin 1 structural; renin b; renin precursor, renal; renin-1; Rn-1; Rnr | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 5972 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.