missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Renin Substrate Tetradecapeptide, Rat

For cellular and molecular biology applications

€ 58.07 - € 158.00

Specifications

Color White to off-white
Form Lyophilized
Molecular Weight (g/mol) 1823.08
Storage Requirements -30° to -10°C
View More Specs

Products 2
Product Code Brand Substrate Quantity Price Quantity & Availability  
Product Code Brand Substrate Quantity Price Quantity & Availability  
15448119
SDS View Documents Promotion Details
Thermo Scientific™
J66671.#0
5mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15458119
SDS View Documents Promotion Details
Thermo Scientific™
J66671.M^
1mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

White to off-white
Lyophilized
1823.08
-30° to -10°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only