missing translation for 'onlineSavingsMsg'
Learn More

Reparixin L-lysine salt, MedChemExpress

Product Code. 16458987
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16458987

Brand: Medchem Express HY15252

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Reparixin L-lysine salt is an allosteric inhibitor of chemokine receptor 1/2 (CXCR1/2) activation.

Reparixin L-lysine salt is an allosteric inhibitor of chemokine receptor 1/2 (CXCR1/2) activation.

Specifications

Chemical Name or Material Reparixin L-lysine salt
CAS 266359-93-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H35N3O5S
Quantity 5 mg
Synonym Repertaxin L-lysine salt
Solubility Information H2O : 100 mg/mL (232.79 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (232.79 mM; Need ultrasonic)
SMILES N[C@@H](CCCCN)C(O)=O.CS(=O)(NC([C@@H](C1=CC=C(CC(C)C)C=C1)C)=O)=O
Molecular Weight (g/mol) 429.57
Formula Weight 429.57
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.