Learn More
SGE™ Repeating Adapter
Click to view available options
:
Repeating adapter can be fitted to manually operated syringes
Description
- Can be preset to any desired volume
- Helps prevent plunger blow out at elevated pressures
- Repeating adapter can be fitted to manually operated syringes
Specifications
Specifications
| For Use With (Application) | Helps prevent plunger blow out at elevated pressures |
| For Use With (Equipment) | Manually Operated Syringe |