Description
This product is specially designed to remove trace level ionic and organic contaminates from EDI, RO or distilled pure water to generate ultrapure water with high resistivity.
Rephile filter pack. Comparable to QGARDT1X1.
Compatible with following Millipore™ systems:
- Milli-Q Advantage A10™
- Milli-Q Reference™
Only compatible with systems manufactured before 2013.
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Water Purification System |
| Type | Cartidge Kit |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.