missing translation for 'onlineSavingsMsg'
Learn More

REPIN1 Antibody, Novus Biologicals™

Product Code. 30228635 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30228635 20 μL 20µL
30226898 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30228635 Supplier Novus Biologicals Supplier No. NBP33351920ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

REPIN1 Monoclonal antibody specifically detects REPIN1 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen REPIN1
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000 - 1:48000, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.3), 50% glycerol, 0.05% BSA
Gene Alias 60 kDa origin-specific DNA-binding protein, 60 kDa replication initiation region protein, ATT-binding protein, DHFR oribeta-binding protein RIP60, H_DJ0584D14.12, replication initiation region protein (60kD), replication initiator 1, RIP60AP4, Zfp464, Zinc finger protein 464, zinc finger protein 464 (RIP60), zinc finger protein AP4, zinc finger proten AP4, ZNF464
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 376-567 of human REPIN1 (NP_037532.2).,, Sequence:, SCDDCGRSFRLERFLRAHQRQHTGERPFTCAECGKNFGKKTHLVAHSRVHSGERPFACEECGRRFSQGSHLAAHRRDHAPDRPFVCPDCGKAFRHKPYLAAHRRIHTGEKPYVCPDCGKAFSQKSNLVSHRRIHTGERPYACPDCDRSFSQKSNLITHRKSHIRDGAFCCAICGQTFDDEERLLAHQKKHDV
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29803
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.