Learn More
Idex Replacement Ferrules for SealTight™ Fittings
Product Code. 11593970 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1/Ea
10/pk
Unit Size:
Each
Pack of 10
Product Code. 11593970
Brand: Idex F192
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Slotted end on ferrule creates second compression zone for a tight seal
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Color Black
Description 10-32 or M6 coned, for 1/16 in. OD tubing
Material PEEK
Quantity 1/Ea