Learn More
Idex Replacement Ferrules for SealTight™ Fittings
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1/Ea
10/pk
Unit Size:
Each
Pack of 10
Specifications
Specifications
| Type | Ferrule |
| Color | Black |
| Description | 10-32 or M6 coned, for 1/16 in. OD tubing |
| Material | PEEK |
| Quantity | 1/Ea |