Learn More
Hamilton™ Replacement Needles for Waters U6K HPLC Injection Valve Syringes
Product Code. 11756287
Click to view available options
Needle Point Style:
Removable Square/Blunt
Product Type:
Large Hub Replacement Needle
Small Hub Replacement Needle
Unit Size:
Pack of 6
Product Code. 11756287
Brand: Hamilton™ 864701
Small and large hub removable replacement needles
  • Standard needle length is 2 in. / 51 mm
  • Available in convenient six packs
  • Custom needle lengths and point styles are available
Specifications
Autoclavable Autoclavable
Needle Gauge 25s G
Length (English) Needle 1.97 in.
Product Type Small Hub Replacement Needle
Needle Point Style Removable Square/Blunt
For Use With (Equipment) Waters U6K HPLC Injection Valve Syringes