Learn More
Merck Accessories for Pressure Filter Holders, 47 mm
Shop All Merck Products
Click to view available options
Type:
Filter Support Screen
O-Ring
Replacement Parts Kit
Material:
PTFE
Stainless Steel
Unit Size:
Each
Pack of 5
Description
Accessories for filter holders for sample preparation.
For use with 47 mm pressure filter holders.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | PTFE |
| Sterility | Non-sterile |
| Includes | (5) Top cap O-Rings, (5) locking ring gasket, (5) filter sealing O-Ring, (5) screen gasket |
| Type | Replacement Parts Kit |