Learn More
Grant Instruments™ Polycarbonate Transparent Lid, Gabled
Click to view available options
For Use With (Equipment):
26L Tanks
2L Tanks
5L Tanks. Fits JBA5/JBN5 bath
SAP12, JBN12, JBA12, SBB Aqua 12+
Förpackningsstorlek:
Each
Description
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
- Polycarbonate construction
- Clear, gabled lid allows condensation buildup to drip away from immersed vessels.
- For use with Sub Aqua Pro Series Unstirred Water Baths
Specifications
Specifications
| Description | Gabled |
| For Use With (Equipment) | SAP12, JBN12, JBA12, SBB Aqua 12+ |
| For Use With (Application) | Used to avoid contamination, reduce evaporation, and save energy |
| Material | Polycarbonate |