Learn More
Duran™ Replacement Sidearm Caps
Product Code. 15820488 Shop All DWK Life Sciences Products
Change view
Click to view available options
:
45 mm size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15820488 45 mm size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15820488 Supplier Duran™ Supplier No. W356872
Replacement caps for spinner flask sidearms.
  • Accessory item
  • Aids in cell culture applications
  • 45 mm size
  • Manufactured from white PBT with a PTFE/silicone liner
  • Provided in packs of 2
TRUSTED_SUSTAINABILITY
Specifications
Closure Color White
Material PBT
For Use With (Equipment) Spinner flasks
Includes PTFE/silicone liner
No. per Pack 2/pk
Closure Type Sidearm cap
Closure Size 45 mm