missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ REPS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29422
This item is not returnable.
View return policy
Description
REPS1 Polyclonal antibody specifically detects REPS1 in Human samples. It is validated for Immunoprecipitation
Specifications
| REPS1 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 85021 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| RALBP1, RALBP1 associated Eps domain containing 1, ralBP1-associated Eps domain-containing protein 1, RalBP1-interacting protein 1 | |
| Between 746 and 796 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction