missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RERE Polyclonal Antibody
GREENER_CHOICE

Product Code. 15524262
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15524262 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15524262 Supplier Invitrogen™ Supplier No. PA540586

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: RPRRSCTLEG GAKNYAESDH SEDEDNDNNS ATAEESTKKN KKKPPKKKSR Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RERE
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RERE
Gene Accession No. Q9P2R6
Gene Alias 1110033A15Rik; AI414665; ARG; arginine glutamic acid dipeptide (RE) repeats; arginine-glutamic acid dipeptide (RE) repeats; arginine-glutamic acid dipeptide repeats; arginine-glutamic acid dipeptide repeats protein; Arp; ATN1L; Atr2; atrophin 2; atrophin-1 like protein; atrophin-1 related protein; Atrophin-1-like protein; Atrophin-1-related protein; atrophin-2; AW742570; DNB1; eye>m03Jus>; eyes3; FLJ38775; KIAA0458; mKIAA0458; NEDBEH; Rere
Gene Symbols RERE
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RERE (aa 36-85).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 473
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.