missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH Reservoir Bottle for Automatic Burettes

Product Code. 10036141
Change view
Click to view available options
Color:
Amber
Clear
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10036141 Clear Each
10466691 Amber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10036141 Supplier Poulten & Graf GmbH Supplier No. 1 06363

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Poulten & Graf GmbH Reservoir Bottle for Automatic Burettes are ideal for titrations.

  • For automatic burrette pellet pattern
  • TS 29, 2/32

Specifications

Color Clear
Capacity (Metric) 2000 mL
For Use With (Equipment) Automatic Burette
Material Glass
No. per Pack 1
Type Reservoir Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.