missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Resistin Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-203JF549
This item is not returnable.
View return policy
Description
Resistin Polyclonal antibody specifically detects Resistin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Resistin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Janelia Fluor 549 | |
| Adipose tissue-specific secretory factor, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN1, RSTNXCP1 | |
| Reacts with residues 79-90 [GSWDVRAETTCH] of the human Resistin protein. | |
| 0.1 mL | |
| Diabetes Research | |
| 56729 | |
| Store at 4°C in the dark. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu