missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Resistin Antibody (MRES18), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80087AF488
This item is not returnable.
View return policy
Description
Resistin Monoclonal antibody specifically detects Resistin in Mouse samples. It is validated for ELISA
Specifications
| Resistin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Adipose tissue-specific secretory factor, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN1, RSTNXCP1 | |
| Recombinant mouse resistin. | |
| 0.1 mL | |
| Diabetes Research | |
| 56729 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| MRES18 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction