Learn More
Macherey-Nagel™ Resolvosil BSA-7 CC Guard Columns Cartridges
Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1 Pack |
| Particle Size | 7 μm |
| Length (Metric) | 8 mm |
| Pore Size | 300Å |
| Type | CC Guard Columns Cartridges |
| Diameter (Metric) Inner | 4 mm |
| For Use With (Equipment) | EC columns of 4 and 4.6 mm ID with guard column adapter EC (REF 721359) |
| Stationary Phase | Resolvosil BSA-7 |