missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus A2 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93588C
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus A2 Polyclonal antibody specifically detects Respiratory Syncytial Virus A2 in Human, Virus samples. It is validated for Western Blot
Specifications
| Respiratory Syncytial Virus A2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Human, Virus | |
| Purified |
| Western Blot | |
| DyLight 650 | |
| Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV | |
| Reacts with residues 180-198 TCWAICKRIPNKKPGKK of the human RSV G protein A2 strain. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction