missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Antibody (9C5cc), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
€ 519.00
Specifications
| Antigen | Respiratory Syncytial Virus |
|---|---|
| Clone | 9C5cc |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Janelia Fluor 646 |
Description
Respiratory Syncytial Virus Monoclonal antibody specifically detects Respiratory Syncytial Virus in Virus samples. It is validated for ELISA, Lateral FlowSpecifications
| Respiratory Syncytial Virus | |
| ELISA | |
| Janelia Fluor 646 | |
| Mouse | |
| Virus | |
| Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV | |
| IgG2b | |
| Protein A purified |
| 9C5cc | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified RSV, strain Long. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title