missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Glycoprotein F Antibody (11-2-C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 351.00
Specifications
| Antigen | Respiratory Syncytial Virus Glycoprotein F |
|---|---|
| Clone | 11-2-C9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Functional |
| Applications | Western Blot, ELISA, Immunofluorescence, Functional Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697917
|
Novus Biologicals
NBP2-50156-0.025ml |
0.025 mL |
€ 190.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604977
|
Novus Biologicals
NBP2-50156 |
0.1 mL |
€ 351.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Respiratory Syncytial Virus Glycoprotein F Monoclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Functional AssaySpecifications
| Respiratory Syncytial Virus Glycoprotein F | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunofluorescence, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Virology Bacteria and Parasites | |
| PBS | |
| IgG1 κ | |
| Protein A purified |
| 11-2-C9 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Gradient-purified RSF-44 virus (subgroup A) UV inactivated for 20 minutes at 20C | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title