missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Glycoprotein F Antibody (11-6-F9), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50394UV
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus Glycoprotein F Monoclonal specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Respiratory Syncytial Virus Glycoprotein F | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Gradient-purified RSF-44 virus (subgroup A) UV inactivated for 20 minutes at 20C | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Immunoblot using the reduced and unreduced RS virus shows that 11-6-F9 reacts with the non-reduced form of the virus F protein (VP70). It has a plaque reduction neutralization titer of 10 1.3. Induce in vitro antibody-dependent enhancement of macrophage cells.This antibody has a RS virus 90% plaque neutralization titre of 101.3.and it also has RSV enhancing activity (enhancement index range: 2.5-5.3). Gimenez et al. (1996). Gimenez et al. (1996) was the first report showing that a monoclonal antibody specific for the F protein has enhancing activity. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 11-6-F9 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction