Learn More
Respiratory Syncytial Virus Glycoprotein F Antibody (342) [HRP], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-43763H
Product Code. 30281326
  • € 561.00 / 0.10mL
Description
Respiratory Syncytial Virus Glycoprotein F Monoclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for ELISA,Lateral Flow
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Respiratory Syncytial Virus Glycoprotein F
Monoclonal
HRP
PBS
Rabbit
Protein A purified
RUO
Virus
Purified
ELISA, Lateral Flow
342
ELISA, Lateral Flow Assay
Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2
This antibody was obtained from a rabbit immunized with purified, recombinant Human Respiratory Syncytial Virus A2 Fusion Protein (Uniprot#: P03420-1; Met 1-Thr 529).
0.1 mL
Primary
Store at 4°C in the dark.