missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Glycoprotein G Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18801F
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus Glycoprotein G Polyclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein G in Virus samples. It is validated for ELISA
Specifications
| Respiratory Syncytial Virus Glycoprotein G | |
| Polyclonal | |
| ELISA | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA | |
| FITC | |
| PBS | |
| Produced in rabbits immunized with purified recombinant Respiratory Syncytial Virus Glycoprotein G (Ser64-Lys321, Accession: AII22117.1) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction