Learn More
Respiratory Syncytial Virus M2-1 Protein Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-93589PCP
Product Code. 18508512
  • € 531.00 / 0.10mL
Description
Respiratory Syncytial Virus M2-1 Protein Polyclonal antibody specifically detects Respiratory Syncytial Virus M2-1 Protein in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rabbit
Reacts with residues 81-95 [LESYIGSINNITKQSA] of the human Respiratory Syncytial Virus M2 protein, which is a CTL epitope. 93% identity to bovine RSV M2.