missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory syncytial virus Mouse anti-Respiratory syncytial virus, Clone: J360, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13640.1mg
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against respiratory syncytial virus (RSV).
- High specificity
- High reactivity
Specifications
| Respiratory syncytial virus | |
| Monoclonal | |
| Unconjugated | |
| ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Respiratory Syncytial virus (RSV) | |
| Liquid |
| ELISA, Immunofluorescence, Lateral Flow | |
| J360 | |
| Mouse monoclonal antibody raised against respiratory syncytial virus (RSV). | |
| In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
| Protein A purification | |
| RUO | |
| This antibody reacts with respiratory syncytial virus types A and B fusion protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction