missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Nucleoprotein Antibody (1C3), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50534V
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus Nucleoprotein Monoclonal specifically detects Respiratory Syncytial Virus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Respiratory Syncytial Virus Nucleoprotein | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| hRSV strain A2 infected HeLa cells | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| This antibody has broad specificity for hRSV of both sub-groups A and B. | |
| Store at 4C in the dark. | |
| IgG2a |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1C3 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction