missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Nucleoprotein Antibody (RSV3132 (B023)), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64752B
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus Nucleoprotein Monoclonal antibody specifically detects Respiratory Syncytial Virus Nucleoprotein in Human, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| Respiratory Syncytial Virus Nucleoprotein | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Bovine RSV strains: 127, SNK and 9007. Human RSV strains: A2, Long, Randall and 8/60. | |
| 0.5 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| RSV3132 (B023) | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction