Learn More
Respiratory Syncytial Virus Nucleoprotein Antibody (RSV3132 (B023)) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-64752
Product Code. 18776403
  • € 765.00 / 0.50mg
Description
Respiratory Syncytial Virus Nucleoprotein Monoclonal specifically detects Respiratory Syncytial Virus Nucleoprotein in Human, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Respiratory Syncytial Virus Nucleoprotein
Monoclonal
1.0 mg/mL
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:100
Bovine RSV strains: 127, SNK and 9007. Human RSV strains: A2, Long, Randall and 8/60.
0.5 mg
Virology Bacteria and Parasites
Recognizes an epitope within the 42kD RSV nucleoprotein.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
RSV3132 (B023)
Unconjugated
Mouse
Protein A or G purified
RUO
Primary
Virus