missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Subgroup A G Glycoprotein Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18799B
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus Subgroup A G Glycoprotein Polyclonal antibody specifically detects Respiratory Syncytial Virus Subgroup A G Glycoprotein in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Respiratory Syncytial Virus Subgroup A G Glycoprotein | |
| Polyclonal | |
| Western Blot, ELISA | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| Biotin | |
| PBS | |
| Produced in rabbits immunized with purified recombinant Human Respiratory Syncytial Virus Subgroup A G Glycoprotein (His67-Gln298, Uniprot: P20895-1) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction