missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REST Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143238
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Transcriptional repressor which binds neuron-restrictive silencer element (NRSE) and represses neuronal gene transcription in non-neuronal cells. Restricts the expression of neuronal genes by associating with two distinct corepressors, mSin3 and CoREST, which in turn recruit histone deacetylase to the promoters of REST-regulated genes. Mediates repression by recruiting the BHC complex at RE1/NRSE sites which acts by deacetylating and demethylating specific sites on histones, thereby acting as a chromatin modifier.
Specifications
| REST | |
| Polyclonal | |
| Unconjugated | |
| REST | |
| 2610008J04Rik; AA407358; Neural-restrictive silencer factor; neuron restrictive silencer factor; neuron-restrictive silencer factor; NRSF; RE1 silencing transcription factor; RE1-silencing transcription factor; RE1-silencing transcription factor variant E1a/E2/E3/E4c; RE1-silencing transcription factor variant E1a/E2/E3/E5; RE1-silencing transcription factor variant E1a/E2/E3/N3a/E4i; RE1-silencing transcription factor variant E1a/E2/E3/N3c/E4; RE1-silencing transcription factor variant E1a/E2/E4; RE1-silencing transcription factor variant E1a/E2/E5; RE1-silencing transcription factor variant E1a/E2a/E2k; RE1-silencing transcription factor variant E1a/E2d/E4g; RE1-silencing transcription factor variant E1a/E2e/E4h; RE1-silencing transcription factor variant E1a/E2f/E4e; RE1-silencing transcription factor variant E1a/E2k/E2i/E3/E4j; RE1-silencing transcription factor variant E1b/E2/E3/E5; RE1-silencing transcription factor variant E1b/E2/E3/N3b/E4i; RE1-silencing transcription factor variant E1b/E2/E3/N3c/E4; RE1-silencing transcription factor variant E1b/E2a/E2k; RE1-silencing transcription factor variant E1b/E2c/E2j/E3/E4; RE1-silencing transcription factor variant E1b/E2e/E4h; RE1-silencing transcription factor variant E1c/E2/E3/E5; RE1-silencing transcription factor variant E1c/E2a/E2k; RE1-silencing transcription factor variant E1c/E2g/E3/E4; repressor binding to the X2 box; Rest; REST4; WT6; X2 box repressor; XBR; zinc finger transcription factor REST protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5978 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q13127 | |
| REST | |
| Peptide with sequence C-PETPGAEDKGKSSK. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction