missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ret Antibody (Regeneron patent anti-RET), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28578-50ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Ret Monoclonal antibody specifically detects Ret in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| Ret | |
| Monoclonal | |
| Unconjugated | |
| P07949 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Regeneron patent anti-RET | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
| c-RET | |
| 50 μg | |
| Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
| 5979 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction