missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ret Antibody (RET/2795), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79897R
This item is not returnable.
View return policy
Description
Ret Monoclonal specifically detects Ret in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ret | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RET | |
| Recombinant fragment (around aa 702-848) of human Ret protein (exact sequence is proprietary) (Uniprot: P07949) | |
| 0.1 mL | |
| Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
| Human | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RET/2795 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction