missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASF1b Antibody (004), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90279C
This item is not returnable.
View return policy
Description
ASF1b Monoclonal specifically detects ASF1b in Human samples. It is validated for Western Blot.
Specifications
| ASF1b | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 4 | |
| Western Blot | |
| ASF1 silencing function 1 homolog B (S. cerevisiae), CCG1-interacting factor A-II, CIA-II, FLJ10604, hAsf1, hAsf1b, hCIA-II, histone chaperone ASF1B, silencing function protein 1 homolog B | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ASF1b (Q9NVP2; Met1-Ile202). | |
| 0.1 mL | |
| Cancer, DNA replication Transcription Translation and Splicing | |
| 55723 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction