missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RET Polyclonal Antibody
GREENER_CHOICE

Product Code. 11592350
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11592350 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11592350 Supplier Invitrogen™ Supplier No. PA517456

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

The receptor tyrosine kinase RET is a member of the cadherin superfamily. RET plays a crucial role in neural crest development, and can undergo oncogenic activation by cytogenetic rearrangement. RET transduce signals for cell growth and differentiation. Mutations in the RET gene are associated with the disorders multiple endocrine neoplasia, type IIA, multiple endocrine neoplasia, type IIB, Hirschsprung disease, and medullary thyroid carcinoma. Two transcript variants encoding different isoforms have been found for the RET gene. Additional transcript variants have been described for RET but their biological validity has not been confirmed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RET
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene RET
Gene Accession No. P07949
Gene Alias Cadherin family member 12; Cadherin family member 12 (CDHF12); Cadherin related family member 16 (CDHR16); cadherin-related family member 16; CDHF12; CDHR16; CG1061; CG14396; CG14396-PA; CG14396-PB; CG14396-PC; c-Ret; CUX1/RET fusion; Dmel\CG14396; Dmel_CG14396; DmHD-59; DmRet; Dret; D-ret; EC 2.7.10.1; ELKS; Extracellular cell-membrane anchored RET cadherin 120 kDa fragment; HD-59; HSCR1; Hydroxyaryl protein kinase; hydroxyaryl-protein kinase; kinase Ret; MEN2; MEN2A; MEN2B; MTC1; Multiple endocrine neoplasia and medullary thyroid carcinoma 1; Oncogene RET; OTTHUMP00000216967; proto-oncogene c-Ret; Proto-oncogene tyrosine-protein kinase receptor Ret; PTC; rearranged during transfection; receptor tyrosine kinase; ret; RET ELE1; Ret gene for receptor tyrosin; Ret oncogene; ret proto-oncogene; ret proto-oncogene (multiple endocrine neoplasia and medullary thyroid carcinoma 1, Hirschsprung disease); Ret proto-oncogene (multiple endocrine neoplasia MEN2A MEN2B and medullary thyroid carcinoma 1 Hirschsprung disease); RET receptor tyrosine kinase; RET transforming sequence; RET51; RET9; RET-ELE1; Reto; Ret-PA; Ret-PB; Ret-PC; Soluble RET kinase fragment
Gene Symbols RET
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Thr1085 of human Ret.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5979
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.