missing translation for 'onlineSavingsMsg'
Learn More

Retaspimycin Hydrochloride, MedChemExpress

Product Code. 16413718
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16413718 10 mg 10mg
16486447 5 mg 5mg
16480097 50 mg 50mg
16454786 100 mg 100mg
16430979 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16413718 Supplier Medchem Express Supplier No. HY10210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Retaspimycin Hydrochloride is a potent inhibitor of Hsp90 with EC50s of 119 nM for both Hsp90 and Grp9.

Retaspimycin Hydrochloride is a potent inhibitor of Hsp90 with EC50s of 119 nM for both Hsp90 and Grp9.

Specifications

Chemical Name or Material Retaspimycin Hydrochloride
CAS 857402-63-2
Color Pink
Purity Grade Notes Research
Molecular Formula C31H46ClN3O8
Quantity 10 mg
Synonym IPI-504
Solubility Information 1M HCl : 100 mg/mL (160.21 mM; ultrasonic and adjust pH to 1 with 1M HCl) ∣DMSO : 60 mg/mL (96.13 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(NC1=C2O)/C(C)=C/C=C\[C@H](OC)[C@H](/C(C)=C/[C@@H]([C@H]([C@H](C[C@@H](CC2=C(C(O)=C1)NCC=C)C)OC)O)C)OC(N)=O.Cl
Molecular Weight (g/mol) 624.17
Formula Weight 624.17
Percent Purity 98.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.