missing translation for 'onlineSavingsMsg'
Learn More

Retinol (All Trans) from synthetic, 99.18%, MP Biomedicals™

Product Code. 11421771 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421771 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11421771 Supplier MP Biomedicals Supplier No. 0219026891

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Retinol is one of the animal forms of vitamin A. It is a diterpenoid and an alcohol

TRUSTED_SUSTAINABILITY

Specifications

CAS 68-26-8
Color Orange
Molecular Formula C20H30O
MDL Number MFCD00001552
Quantity 1 g
Synonym retinol, vitamin a, all-trans-retinol, vitamin a1, alphalin, axerophthol, vitamin a alcohol, oleovitamin a, chocola a, alphasterol
InChI Key FPIPGXGPPPQFEQ-OVSJKPMPSA-N
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCO)C)C
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraen-1-ol
Molecular Weight (g/mol) 286.459
PubChem CID 445354
ChEBI CHEBI:17336
Formula Weight 286.5
Physical Form Powder
Chemical Name or Material Retinol (All Trans), from Synthetic; For HPLC analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • All Trans Retinol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H360-May damage fertility or the unborn child.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: retinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.