missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Retinol Binding Protein RBP Antibody (SPM442), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-34410CL1
This item is not returnable.
View return policy
Description
Retinol Binding Protein RBP Monoclonal antibody specifically detects Retinol Binding Protein RBP in Goat,Human,Monkey,Mouse,Rabbit,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Retinol Binding Protein RBP | |
| Monoclonal | |
| CoraFluor 1 | |
| C, Cellular retinol-binding protein, CRABP-I, CRBP1CRBP-I, CRBPCellular retinol-binding protein I, CRBPI, RBPC, retinol binding protein 1, cellular, retinol-binding protein 1, retinol-binding protein 1, cellular | |
| Retinol binding protein purified from human plasma | |
| 0.1 mL | |
| Cancer, Diabetes Research, Lipid and Metabolism | |
| 5947 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| SPM442 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Goat, Human, Monkey, Mouse, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction