missing translation for 'onlineSavingsMsg'
Learn More

RETN Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen RETN
Concentration 250 μg/mL
Applications Immunohistochemistry, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236112
View Documents
Proteintech
18170-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30235411
View Documents
Proteintech
18170-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The RETN antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human RETN. This antibody recognizes human antigen. The RETN antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications

Specifications

RETN
Immunohistochemistry, ELISA
Unconjugated
Rabbit
Human
ADSF; FIZZ3; MGC126603; MGC126609; RETN1; RSTN; XCP1
IgG
Antigen affinity purification
Polyclonal Antibody
250 μg/mL
Polyclonal
Liquid
RUO
56729
Fusion Protein
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.