Learn More
Brady™ Retractable Barrier Belt with Black Posts
Brand:  Brady™ 912657
Additional Details : Weight : 12.00000kg
 View more versions of this product
Product Code. 11751136
  • € 439.00 / Each
Estimated Shipment
23-02-2021
Specifications
1Pack
Black
100mm
2m
12kg
Retractable Barrier Belt
Retractable Barrier Belt
Retractable Barrier Belt - 2 Metres with Black Posts