Learn More
Brady™ Retractable Barrier Belt with Silver Posts
Brand:  Brady™ 912675
Additional Details : Weight : 12.00000kg
 View more versions of this product
Product Code. 11731156
  • € 439.00 / Each
Estimated Shipment
04-03-2021
Specifications
1Pack
Green
100mm
2m
12kg
Retractable Barrier Belt
Retractable Barrier Belt
Retractable Barrier Belt - 2 Metres with Silver Posts
Silver