Learn More
Brady™ Retro Reflective All Weather Label Tape for M6/M7 Printers
Highly reflective material shines brightly when struck by light which is ideal for applications where high visibility is critical.
Brand: Brady™ 174130
This item is not returnable.
View return policy
Description
- Ideal for labeling job site tools, hard hats, utility enclosures, poles and pedestals, rooftop equipment, racks and shelves
- Retroreflective sheeting and pressure sensitive acrylic adhesive meet Federal Spec L-S-300 for Reflectivity
- Average outdoor durability is 4 to 6 years
- Service temperature of -40°C to 70°C (-40°F to 158°F)
- Made of B-584 plastic material
Specifications
| Not Autoclavable | |
| Permanent Acrylic | |
| Yellow | |
| Continuous | |
| Single-side Printable | |
| Label | |
| Component and Equipment Labeling, Facility Identification, General Identification, Safety Identification | |
| No | |
| Rectangle | |
| -40°C to +70°C | |
| 0.14 mm | |
| 30 ft. | |
| 2 in. | |
| M6C-2000-584-YL |
| ASTM D 2979, Fed. Spec. L-S-300 (Class 1) | |
| None | |
| Gloss | |
| Plastic | |
| Thermal Transfer | |
| Non-sterile | |
| M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | |
| 30 ft. | |
| -40°F to +158°F | |
| 0.0055 in. | |
| 30 ft. x 2 in. | |
| 9.14 m | |
| 50.8 mm |