Learn More
RETSCH PM 100 Planterary Ball Mill Grinding Balls
Product Code. 10324531 Shop All Retsch Products
Change view
Click to view available options
Material:
Agate
Stainless Steel
Zirconium Oxide
Diameter (Metric):
0.2 cm
0.3 cm
1 cm
2 cm
For Use With (Application):
Ideal for colloidal milling
Ideal for standard grinding
Unit Size:
500g
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Diameter (Metric) For Use With (Application) unitSize
10324531 Agate 2 cm Ideal for standard grinding Each
11797044 Stainless Steel 0.3 cm Ideal for colloidal milling 500g
11301460 Stainless Steel 0.2 cm Ideal for colloidal milling Each
10289201 Zirconium Oxide 1 cm Ideal for standard grinding Each
10491213 Stainless Steel 1 cm Ideal for colloidal milling Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10324531 Supplier RETSCH Supplier No. 05.368.0028
Only null left
Add to basket
Add to basket
For use with Retsch PM 100 and PM 200 Planetary Ball Mills
  • In assorted diameters and materials
  • Smaller balls offered for colloidal milling
Specifications
Product Type Grinding Ball
Diameter (English) 0.78 in.
Diameter (Metric) 2 cm
For Use With (Application) Ideal for standard grinding
For Use With (Equipment) PM 100 and PM 200 planetary ball mill
Material Agate