Description
- Accessory for use with ZM 200 Ultra Centrifugal Mill
- Wear resistant coating
Specifications
Specifications
| Material | Stainless Steel |
| Type | 6 Tooth Rotor |
| Max. Speed | 6,000 to 18,000 rpm |
| Max. Tube Diameter | 9.9 cm |
| For Use With (Equipment) | Centrifugal Mill |