Learn More
RETSCH ZM200 Ultra Centrifugal Mill: Distance Sieves
Product Code. 11904252 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
0.01 in. (0.50 mm)
0.08 mm
0.12 mm
0.20 mm
0.25 mm
0.75 mm
1.50 mm
4.0 mm
Pore Style:
Conidur holes
Round holes
Trapezoid holes
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Pore Style Unit Size
11904252 0.08 mm Trapezoid holes Each
10852061 0.12 mm Trapezoid holes Each
10564215 0.20 mm Trapezoid holes Each
10509772 0.75 mm Trapezoid holes Each
10061222 0.25 mm Trapezoid holes Each
10425052 0.01 in. (0.50 mm) Conidur holes Each
10052212 1.50 mm Trapezoid holes Each
11914252 4.0 mm Round holes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11904252 Supplier RETSCH Supplier No. 03.647.0253
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For temperature-sensitive materials
Specifications
Product Type Distance Sieve
For Use With (Equipment) ZM200 ultra centrifugal mil
Material Stainless Steel
Pore Size 0.08 mm
Pore Style Trapezoid holes