Learn More
Fisherbrand™ Reusable Dispenser Tips
Robust, stable, and durable dispenser tips for several brands of pipettors.
Brand:  Fisherbrand™
Additional Details : Weight : 0.05000kg
Product Code. 10010311
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
These tips are for use with the following pipettors:
  • Brand Handystep™ and Handystep™ electronic All tips
  • Eppendorf™ Multipette™ (not Multipette™ Plus) All tips
  • Gilson Distriman™ 
  • Ritter Ripette™ All tips
  • Tri-continent Easystep™ 
Additonal features:
  • These Dispenser™ tips are non-sterile and available by gamma irradiation
  • These dispenser tips are re-useable
  • The adapter for this dispenser tip is autoclavable
Specifications
Specifications
With one autoclavable adapter
50 mL
Brand Handystep and Handystep, Electronic Eppendorf, Multipette (not Multipette Plus), Gilson Distriman, Ritter Ripette. Tri-continent Easystep
25/Pk.
Dispenser Tip